accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64808-48-6

IUPAC Name: disodium 2-[(2-carboxylatophenyl)amino]-4-chlorobenzoate
Synonyms: lobenzarit
Molecular Weight (g/mol): 335.65
Molecular Formula: C14H8ClNNa2O4
InChi Key: QPJBONAWFAURGB-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1NC1=CC(Cl)=CC=C1C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results