accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64913-16-2

IUPAC Name: 4,5,6-tris(acetyloxy)-2-methyloxan-3-yl acetate
Synonyms: 4,5,6-tris(acetyloxy)-2-methyloxan-3-yl acetate
Molecular Weight (g/mol): 332.31
Molecular Formula: C14H20O9
InChi Key: QZQMGQQOGJIDKJ-UHFFFAOYNA-N
SMILES: CC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results