accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6494-19-5

IUPAC Name: 3-methyl-6-nitro-2H-indazole
Synonyms: 3-methyl-6-nitro-2H-indazole
Molecular Weight (g/mol): 177.16
Molecular Formula: C8H7N3O2
InChi Key: FUNWSYKLFDLUIZ-UHFFFAOYSA-N
SMILES: CC1=C2C=CC(=CC2=NN1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results