accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COClCH3N

CAS RN 64951-58-2

IUPAC Name: 4-chloro-8-methoxy-2-methylquinoline
Synonyms: 4-chloro-8-methoxy-2-methylquinoline
Molecular Weight (g/mol): 207.66
Molecular Formula: C11H10ClNO
InChi Key: RHSCPZBPNUZBOA-UHFFFAOYSA-N
SMILES: COC1=C2N=C(C)C=C(Cl)C2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results