accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 649736-31-2

CH3OFHONOO

CAS RN 649736-31-2

IUPAC Name: 1-(2-fluoro-3-hydroxy-6-nitrophenyl)propan-2-one
Synonyms: 1-(2-fluoro-3-hydroxy-6-nitrophenyl)propan-2-one
Molecular Weight (g/mol): 213.16
Molecular Formula: C9H8FNO4
InChi Key: IPRZVDNMKMONNN-UHFFFAOYSA-N
SMILES: CC(=O)CC1=C(C=CC(O)=C1F)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results