accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65032-66-8

IUPAC Name: 2-(2-methyl-1,3-thiazol-4-yl)benzoic acid
Synonyms: 2-(2-methyl-1,3-thiazol-4-yl)benzoic acid
Molecular Weight (g/mol): 219.26
Molecular Formula: C11H9NO2S
InChi Key: BNRSCIXYHUTATP-UHFFFAOYSA-N
SMILES: CC1=NC(=CS1)C1=CC=CC=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results