accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65036-65-9

IUPAC Name: sodium 4-(2-bromoethyl)benzene-1-sulfonate
Synonyms: sodium 4-(2-bromoethyl)benzenesulfonate
Molecular Weight (g/mol): 287.1
Molecular Formula: C8H8BrNaO3S
InChi Key: ZFGDZMNLHTUEAV-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CC=C(CCBr)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results