accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOONN(S)HNOOCH3H3CCH3OHOHN

CAS RN 65057-34-3

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-[1-(4-methylbenzenesulfonyl)-1H-imidazol-4-yl]propanoic acid; N-cyclohexylcyclohexanamine
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-[1-(4-methylbenzenesulfonyl)imidazol-4-yl]propanoic acid; dicha
Molecular Weight (g/mol): 590.78
Molecular Formula: C30H46N4O6S
InChi Key: RNRUVXUHYUOWJW-RSAXXLAASA-N
SMILES: C1CCC(CC1)NC1CCCCC1.CC1=CC=C(C=C1)S(=O)(=O)N1C=NC(C[C@H](NC(=O)OC(C)(C)C)C(O)=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)