accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH2N

CAS RN 65079-19-8

IUPAC Name: 2-methylquinolin-6-amine
Synonyms: 2-methylquinolin-6-amine
Molecular Weight (g/mol): 158.2
Molecular Formula: C10H10N2
InChi Key: TYJFYUVDUUACKX-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=C(N)C=CC2=N1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)