accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFOS

CAS RN 651-70-7

IUPAC Name: 2,2,2-trifluoro-1-(thiophen-2-yl)ethan-1-one
Synonyms: 2,2,2-trifluoro-1-(thiophen-2-yl)ethanone
Molecular Weight (g/mol): 180.14
Molecular Formula: C6H3F3OS
InChi Key: CZYKJGCKVBXLGF-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)C1=CC=CS1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)