accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFHSFFF

CAS RN 651-84-3

IUPAC Name: 2,3,5,6-tetrafluoro-4-(trifluoromethyl)benzene-1-thiol
Synonyms: 2,3,5,6-tetrafluoro-4-(trifluoromethyl)benzenethiol
Molecular Weight (g/mol): 250.13
Molecular Formula: C7HF7S
InChi Key: BXMOMKVOHOSVJH-UHFFFAOYSA-N
SMILES: FC1=C(F)C(=C(F)C(F)=C1S)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results