accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65145-13-3

IUPAC Name: 2-fluoro-4-hydroxybenzoic acid
Synonyms: 2-fluoro-4-hydroxybenzoic acid
Molecular Weight (g/mol): 156.11
Molecular Formula: C7H5FO3
InChi Key: NXWTWYULZRDBSA-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C=C(O)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)