accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65162-13-2

IUPAC Name: 1-methoxy-5-methylphenazin-5-ium methyl sulfate
Synonyms: 1-methoxy-5-methylphenazin-5-ium methyl sulfate(1-)
Molecular Weight (g/mol): 336.36
Molecular Formula: C15H16N2O5S
InChi Key: MASUWVVNWALEEM-UHFFFAOYSA-M
SMILES: COS([O-])(=O)=O.COC1=C2N=C3C=CC=CC3=[N+](C)C2=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results