accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 651780-02-8

IUPAC Name: tert-butyl 5-bromo-1H-indazole-1-carboxylate
Synonyms: tert-butyl 5-bromoindazole-1-carboxylate
Molecular Weight (g/mol): 297.15
Molecular Formula: C12H13BrN2O2
InChi Key: CAQGDWLRIQCXSP-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1N=CC2=CC(Br)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results