accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaCH3NONCH3NNOOO

CAS RN 652-37-9

IUPAC Name: sodium 2-(1,3-dimethyl-2,6-dioxo-2,3,6,7-tetrahydro-1H-purin-7-yl)acetate
Synonyms: acefylline
Molecular Weight (g/mol): 260.19
Molecular Formula: C9H9N4NaO4
InChi Key: MSFVZSOKOXZSME-UHFFFAOYSA-M
SMILES: [Na+].CN1C2=C(N(CC([O-])=O)C=N2)C(=O)N(C)C1=O
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (1)
  • (5)
  • (2)
  • (2)
  • (2)
  • (1)