accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 652-67-5

IUPAC Name: (3R,3aR,6S,6aR)-hexahydrofuro[3,2-b]furan-3,6-diol
Synonyms: isosorbide isosorbidum dianhydrosorbitol devicoran isobide vascardin dinitrate sorbid isosorbide preparation isosorbide (product) isosorbida
Molecular Weight (g/mol): 146.14
Molecular Formula: C6H10O4
InChi Key: KLDXJTOLSGUMSJ-JGWLITMVSA-N
SMILES: O[C@H]1CO[C@@H]2[C@H](O)CO[C@H]12
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (7)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (3)
  • (7)
  • (3)
  • (2)
  • (7)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (7)