CAS RN 6520-83-8
CAS RN 6520-83-8
IUPAC Name:
ethyl 2-methoxy-6-methylbenzoate
Synonyms:
ethyl 2-methoxy-6-methylbenzoate
Molecular Weight (g/mol):
194.23
Molecular Formula:
C11H14O3
InChi Key:
GQLSLEBNODXANJ-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=C(C)C=CC=C1OC
1
–
1
of
1
results
1
Ethyl 2-Methoxy-6-methylbenzoate 95.0+%, TCI America™
CAS: 6520-83-8 Molecular Formula: C11H14O3 Molecular Weight (g/mol): 194.23 MDL Number: MFCD00191666 InChI Key: GQLSLEBNODXANJ-UHFFFAOYSA-N Synonym: 2-Methoxy-6-methylbenzoic Acid Ethyl Ester PubChem CID: 591060 IUPAC Name: ethyl 2-methoxy-6-methylbenzoate SMILES: CCOC(=O)C1=C(C=CC=C1OC)C