accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOH

CAS RN 6521-29-5

IUPAC Name: pentyl 4-hydroxybenzoate
Synonyms: pentyl 4-hydroxybenzoate
Molecular Weight (g/mol): 208.26
Molecular Formula: C12H16O3
InChi Key: ZNSSPLQZSUWFJT-UHFFFAOYSA-N
SMILES: CCCCCOC(=O)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)