accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 652148-91-9

IUPAC Name: (5-chloropyridin-2-yl)boronic acid
Synonyms: 5-chloropyridin-2-ylboronic acid
Molecular Weight (g/mol): 157.36
Molecular Formula: C5H5BClNO2
InChi Key: JEGHCYRKSUGHJH-UHFFFAOYSA-N
SMILES: OB(O)C1=NC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results