accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OO(R)(R)OHOOOCH3H3CCH3OOH

CAS RN 65259-81-6

IUPAC Name: (2R,3R)-2,3-bis[(2,2-dimethylpropanoyl)oxy]butanedioic acid
Synonyms: (2R,3R)-2,3-bis[(2,2-dimethylpropanoyl)oxy]butanedioic acid
Molecular Weight (g/mol): 318.32
Molecular Formula: C14H22O8
InChi Key: UFHJEZDFEHUYCR-YZYOREDDNA-N
SMILES: CC(C)(C)C(=O)O[C@H]([C@@H](OC(=O)C(C)(C)C)C(O)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results