accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6528-85-4

IUPAC Name: 2-(2-methoxyphenyl)-1H-1,3-benzodiazole
Synonyms: 2-(2-methoxyphenyl)-1H-1,3-benzodiazole
Molecular Weight (g/mol): 224.26
Molecular Formula: C14H12N2O
InChi Key: JNFBDHNAZPBWTF-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C1=NC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results