accessibility menu, dialog, popup

UserName

Viewing profile as user:
OFBr

CAS RN 65295-58-1

IUPAC Name: 1-(bromomethyl)-3-(4-fluorophenoxy)benzene
Synonyms: 1-(bromomethyl)-3-(4-fluorophenoxy)benzene
Molecular Weight (g/mol): 281.12
Molecular Formula: C13H10BrFO
InChi Key: JGFSTQUUDSBQCO-UHFFFAOYSA-N
SMILES: FC1=CC=C(OC2=CC=CC(CBr)=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results