CAS RN 65295-61-6
IUPAC Name:
3-(4-fluorophenoxy)benzaldehyde
Synonyms:
3-(4-fluorophenoxy)benzaldehyde
Molecular Weight (g/mol):
216.21
Molecular Formula:
C13H9FO2
InChi Key:
HTBZPEDBMOXWID-UHFFFAOYSA-N
SMILES:
FC1=CC=C(OC2=CC=CC(C=O)=C2)C=C1
1
–
1
of
1
results
1
3-(4-Fluorophenoxy)benzaldehyde, 97%, Thermo Scientific™
CAS: 65295-61-6 Molecular Formula: C13H9FO2 Molecular Weight (g/mol): 216.211 MDL Number: MFCD02258991 InChI Key: HTBZPEDBMOXWID-UHFFFAOYSA-N PubChem CID: 3085320 IUPAC Name: 3-(4-fluorophenoxy)benzaldehyde SMILES: C1=CC(=CC(=C1)OC2=CC=C(C=C2)F)C=O