accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 653-30-5

IUPAC Name: 2-(2,3,4,5,6-pentafluorophenyl)acetonitrile
Synonyms: 2-(2,3,4,5,6-pentafluorophenyl)acetonitrile
Molecular Weight (g/mol): 207.1
Molecular Formula: C8H2F5N
InChi Key: YDNOJUAQBFXZCR-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(CC#N)C(F)=C1F
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)