accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 653-63-4

IUPAC Name: {[(2R,3S,5R)-5-(6-amino-9H-purin-9-yl)-3-hydroxyoxolan-2-yl]methoxy}phosphonic acid
Synonyms: DAMP 2'-deoxy-AMP deoxyadenosine 5'-phosphate damp cpd PdA deoxy-AMP 2'deoxy-5'-AMP deoxyadenosine-phosphate 2'-dAMP deoxyadenosine monophosphate
Molecular Weight (g/mol): 331.23
Molecular Formula: C10H14N5O6P
InChi Key: KHWCHTKSEGGWEX-RRKCRQDMSA-N
SMILES: NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](COP(O)(O)=O)O3)C2=NC=N1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results