accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6531-35-7

IUPAC Name: 2,3-dimethyl-9,10-dihydroanthracene-9,10-dione
Synonyms: 2,3-dimethylanthracene-9,10-dione
Molecular Weight (g/mol): 236.27
Molecular Formula: C16H12O2
InChi Key: KIJPZYXCIHZVGP-UHFFFAOYSA-N
SMILES: CC1=CC2=C(C=C1C)C(=O)C1=CC=CC=C1C2=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results