accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH

CAS RN 6531-86-8

IUPAC Name: [1,1'-bi(cyclohexane)]-2-ol
Synonyms: [1,1'-bi(cyclohexane)]-2-ol
Molecular Weight (g/mol): 182.31
Molecular Formula: C12H22O
InChi Key: YTWWBKKPWACPPB-UHFFFAOYNA-N
SMILES: OC1CCCCC1C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results