accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65347-55-9

IUPAC Name: 3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole
Synonyms: 3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole
Molecular Weight (g/mol): 198.27
Molecular Formula: C13H14N2
InChi Key: CIRSPTXGPFAXRE-UHFFFAOYSA-N
SMILES: C1CC(=CCN1)C1=CNC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results