accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHO

CAS RN 65355-29-5

IUPAC Name: 4-(4-propylcyclohexyl)benzoic acid
Synonyms: 4-(4-propylcyclohexyl)benzoic acid
Molecular Weight (g/mol): 246.35
Molecular Formula: C16H22O2
InChi Key: VACLULPMEXHBMD-UHFFFAOYSA-N
SMILES: CCCC1CCC(CC1)C1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results