accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCH3H3C(R)NH2HOOO(S)OH(R)H2NH

CAS RN 65391-42-6

IUPAC Name: hydrogen (2R,4S,5R)-2,5-diamino-4-hydroxy-2-(2-methylpropyl)-3-oxo-6-phenylhexanoic acid chloride
Synonyms: hydrogen (2R,4S,5R)-2,5-diamino-4-hydroxy-2-(2-methylpropyl)-3-oxo-6-phenylhexanoic acid chloride
Molecular Weight (g/mol): 344.84
Molecular Formula: C16H25ClN2O4
InChi Key: JQOATSUOLWQWKR-KHUBKBNHSA-N
SMILES: [H+].[Cl-].CC(C)C[C@](N)(C(O)=O)C(=O)[C@@H](O)[C@H](N)CC1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)