accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OO

CAS RN 65399-05-5

IUPAC Name: 5-amino-1,3-dihydro-2-benzofuran-1-one
Synonyms: 5-amino-3H-2-benzofuran-1-one
Molecular Weight (g/mol): 149.15
Molecular Formula: C8H7NO2
InChi Key: ISMUWQMUWFPFBZ-UHFFFAOYSA-N
SMILES: NC1=CC=C2C(=O)OCC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)