accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHH3CHO

CAS RN 654-42-2

IUPAC Name: 2,6-dimethylbenzene-1,4-diol
Synonyms: 1,4-benzenediol, 2,6-dimethyl-
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: SGWZVZZVXOJRAQ-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC(C)=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results