accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3CH2H2CCH2

CAS RN 65405-58-5

IUPAC Name: 1,2-diethenylbenzene; [(2-ethenylphenyl)methyl]dimethylamine
Synonyms: [(2-ethenylphenyl)methyl]dimethylamine; divinylbenzene
Molecular Weight (g/mol): 291.44
Molecular Formula: C21H25N
InChi Key: OTCJDTFYONDHFM-UHFFFAOYSA-N
SMILES: C=CC1=CC=CC=C1C=C.CN(C)CC1=CC=CC=C1C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)