accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO(E)

CAS RN 65405-70-1

IUPAC Name: (4E)-dec-4-enal
Synonyms: trans-4-decenal
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: CWRKZMLUDFBPAO-VOTSOKGWSA-N
SMILES: CCCCC\C=C\CCC=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results