accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3SiH3CCH3OBr

CAS RN 65423-56-5

IUPAC Name: (3-bromophenoxy)(tert-butyl)dimethylsilane
Synonyms: 3-bromophenoxy(tert-butyl)dimethylsilane
Molecular Weight (g/mol): 287.27
Molecular Formula: C12H19BrOSi
InChi Key: BFRHMVJJJGUHDI-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OC1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results