accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65473-13-4

IUPAC Name: methyl[(naphthalen-1-yl)methyl]azanium
Synonyms: methyl(naphthalen-1-ylmethyl)azanium
Molecular Weight (g/mol): 172.25
Molecular Formula: C12H14N
InChi Key: MQRIUFVBEVFILS-UHFFFAOYSA-O
SMILES: C[NH2+]CC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)