accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 655-32-3

IUPAC Name: 2,2,2-trifluoro-1-(4-fluorophenyl)ethan-1-one
Synonyms: 2,2,2-trifluoro-1-(4-fluorophenyl)ethanone
Molecular Weight (g/mol): 192.11
Molecular Formula: C8H4F4O
InChi Key: LUKLMXJAEKXROG-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)C(F)(F)F
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)