accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6553-96-4

IUPAC Name: 2,4,6-tris(propan-2-yl)benzene-1-sulfonyl chloride
Synonyms: 2,4,6-triisopropylbenzenesulfonyl chloride
Molecular Weight (g/mol): 302.86
Molecular Formula: C15H23ClO2S
InChi Key: JAPYIBBSTJFDAK-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (4)
  • (3)
Quantity 
  • (3)
  • (4)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (4)
  • (3)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)