accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6554-73-0

IUPAC Name: N-tert-butyl-2-methylprop-2-enamide
Synonyms: N-tert-butyl-2-methylprop-2-enamide
Molecular Weight (g/mol): 141.21
Molecular Formula: C8H15NO
InChi Key: QQZXAODFGRZKJT-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)NC(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results