accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6554-98-9

IUPAC Name: 4-[(1E)-2-phenylethenyl]phenol
Synonyms: trans-4-hydroxystilbene p-styrylphenol
Molecular Weight (g/mol): 196.25
Molecular Formula: C14H12O
InChi Key: QVLMUEOXQBUPAH-VOTSOKGWSA-N
SMILES: OC1=CC=C(\C=C\C2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results