accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2NCH3H3CCH3

CAS RN 65542-26-9

IUPAC Name: [(4-tert-butylphenyl)methyl](methyl)azanium
Synonyms: [(4-tert-butylphenyl)methyl](methyl)azanium
Molecular Weight (g/mol): 178.3
Molecular Formula: C12H20N
InChi Key: ZSHCHOYJMLEAOX-UHFFFAOYSA-O
SMILES: C[NH2+]CC1=CC=C(C=C1)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results