accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NBrI

CAS RN 65550-78-9

IUPAC Name: 2-bromo-5-iodo-3-methylpyridine
Synonyms: 2-bromo-5-iodo-3-methylpyridine
Molecular Weight (g/mol): 297.92
Molecular Formula: C6H5BrIN
InChi Key: WJPSANINUBHCFE-UHFFFAOYSA-N
SMILES: CC1=CC(I)=CN=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results