accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65556-30-1

IUPAC Name: [6-(acetyloxy)-3,4,5-tris(benzyloxy)oxan-2-yl]methyl acetate
Synonyms: [6-(acetyloxy)-3,4,5-tris(benzyloxy)oxan-2-yl]methyl acetate
Molecular Weight (g/mol): 534.61
Molecular Formula: C31H34O8
InChi Key: IFCAMEQHKHEHBS-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(OC(C)=O)C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results