accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNHN

CAS RN 65558-69-2

IUPAC Name: 1-imino-1H-benzo[f]isoindol-3-amine
Synonyms: 3-iminobenzo[f]isoindol-1-amine
Molecular Weight (g/mol): 195.23
Molecular Formula: C12H9N3
InChi Key: JAWNWEKHDFBPSG-UHFFFAOYSA-N
SMILES: NC1=NC(=N)C2=C1C=C1C=CC=CC1=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results