accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHNO

CAS RN 65564-05-8

IUPAC Name: benzyl N-(3-oxopropyl)carbamate
Synonyms: benzyl N-(3-oxopropyl)carbamate
Molecular Weight (g/mol): 207.23
Molecular Formula: C11H13NO3
InChi Key: PQMOZOQTXKMYSK-UHFFFAOYSA-N
SMILES: O=CCCNC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)