CAS RN 65567-06-8
IUPAC Name:
lithiodiphenylphosphane
Synonyms:
lithiodiphenylphosphane
Molecular Weight (g/mol):
192.13
Molecular Formula:
C12H10LiP
InChi Key:
WKUYEGHEUWHKIU-UHFFFAOYSA-N
SMILES:
[Li]P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
2
of
2
results
1
Promotions Available
Lithium diphenylphosphide, 0.5M solution in THF, AcroSeal™