accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 656-35-9

IUPAC Name: 2-(2,4-difluorophenyl)acetonitrile
Synonyms: 2-(2,4-difluorophenyl)acetonitrile
Molecular Weight (g/mol): 153.13
Molecular Formula: C8H5F2N
InChi Key: AGAOESUOSOGZOD-UHFFFAOYSA-N
SMILES: FC1=CC=C(CC#N)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results