accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 656-42-8

IUPAC Name: 2,2-difluoro-2H-1,3-benzodioxole-5-carbaldehyde
Synonyms: 2,2-difluoro-1,3-benzodioxole-5-carbaldehyde
Molecular Weight (g/mol): 186.11
Molecular Formula: C8H4F2O3
InChi Key: GGERGLKEDUUSAP-UHFFFAOYSA-N
SMILES: FC1(F)OC2=CC=C(C=O)C=C2O1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)