accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOFFO

CAS RN 656-46-2

IUPAC Name: 2,2-difluoro-2H-1,3-benzodioxole-5-carboxylate
Synonyms: 2,2-difluoro-1,3-benzodioxole-5-carboxylate
Molecular Weight (g/mol): 201.11
Molecular Formula: C8H3F2O4
InChi Key: VJLDRFCNFNQTTH-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C2OC(F)(F)OC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)