accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 656-57-5

IUPAC Name: 1-bromo-2-fluorocyclohexane
Synonyms: 1-bromo-2-fluorocyclohexane
Molecular Weight (g/mol): 181.05
Molecular Formula: C6H10BrF
InChi Key: AZQRVGXSORXOCR-UHFFFAOYNA-N
SMILES: FC1CCCCC1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results